Anti-TBL1X Rabbit Polyclonal Antibody

Supplier: Genetex
GTX107736
89355-108EA 939.18 CAD
89355-108
Anti-TBL1X Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to TBL1X (transducin (beta)-like 1X-linked)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: TBL1X
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR