Anti-SDCCAG3 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX106180
89354-526EA 939.18 CAD
89354-526
Anti-SDCCAG3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to SDCCAG3 (serologically defined colon cancer antigen 3)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SDCCAG3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR