Anti-OGDH Rabbit Polyclonal Antibody

Supplier: Genetex
GTX105124
89354-118EA 962.63 CAD
89354-118
Anti-OGDH Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to OGDH (oxoglutarate (alpha-ketoglutarate) dehydrogenase (lipoamide))

Suggested starting dilutions are as follows: WB: 1:500-1:10000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: OGDH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR