Anti-IgA Goat Polyclonal Antibody

Supplier: Genetex
GTX77225
89353-552EA 341.07 CAD
89353-552
Anti-IgA Goat Polyclonal Antibody
Antibodies
The affinity purified antibodies react only with IgA by ELISA, immunodiffusion and IEP techniques.. Goats were immunized with highly purified mouse IgA. The resulting antiserum was solid phase absorbed to make it alpha chain specific. The anti alpha was immunoaffinity purified off an IgA-containing solid phase absorbant

Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR