Anti-6xHis Goat Polyclonal Antibody

Supplier: Genetex
GTX30500
89352-664EA 1054.9 CAD
89352-664
Anti-6xHis Goat Polyclonal Antibody
Antibodies
This antibody recognises proteins containing the motif H-H-H-H-H-H and is therefore of value in detecting proteins containing histidine tags. The antibody may be used to track the purification of proteins expressed in bacteria, yeast or insect cells.

Type: Primary
Antigen: 6xHis
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR