Anti-HIF1A Mouse Monoclonal Antibody [clone: H1alpha67]

Supplier: Genetex
GTX30105
89352-644EA 1176.88 CAD
89352-644
Anti-HIF1A Mouse Monoclonal Antibody [clone: H1alpha67]
Antibodies
Mouse monoclonal [H1alpha67] to Hypoxia Inducible Factor-1 alpha

Recommended Starting Dilutions:For WB: Use at a dilution of 1:500.For ChIP: Use at a dilution of 1:10-1:500.For Gel Super Shift Assays: Use at a dilution of 1:1-1:100For ICC/IF:Use at a dilution of 1:50.For IHC:Use at a dilution of 1:20-1:50.For IHC-Fr: Use at a dilution of 1:20-1:50.For IHC-P: Use at a dilution of 1:20-1:50For IP: Use at a dilution of 1:10-1:500.Not tested in other applications. Optimal dilutions/cancentrations should be determined by researcher.

Type: Primary
Antigen: HIF1A
Clonality: Monoclonal
Clone: H1More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR