F1-ClipTip™ Variable Volume Single Channel Pipettes, Thermo Scientific
Supplier: Thermo Fisher Scientific
4641180N 4641210N 4641310N 4641320N
89348-063EA 803.62 CAD
89348-063 89348-069 76299-426 76299-428
F1-ClipTip™ Variable Volume Single Channel Pipettes, Thermo Scientific
Pipettes Micropipettes
Now silent and effortless, Thermo Scientific™ F1-ClipTip™ Variable Volume Pipettes feature newly enhanced volume adjustment, and the added confidence of ClipTip technology that locks tips firmly to the pipette with a complete seal.
  • Enhanced volume adjustment
  • Complete seal
  • Remains sealed until ejection
  • High-quality results
  • More efficient research
  • Lightweight design
  • Secure volume lock
With a light touch, each tip is clipped securely on the pipette. When ClipTip pipette tips are on, they are sealed and will not loosen. The ClipTip pipetting systemMore Product Information