ClipTip® Pipette Tips, 200, Thermo Scientific
Supplier: Thermo Fisher Scientific
94410313 94410310 94410313 94420313 94410318 94420318 94410317
89173-436CS 480.89 CAD
89173-436 89173-466 89348-004 89348-014 89348-024 89348-044 89348-054 89348-034
ClipTip® Pipette Tips, 200, Thermo Scientific
Pipette Tips
ClipTip® pipette tips provide security with a unique and innovative interlocking technology that ensures a complete seal on every channel with minimal tip attachment and ejection force.
All ClipTip® pipette tips feature a hydrophobic inner surface which effectively prevent liquid retention. ClipTip® tips are certified free of RNase, DNase, DNA, ATP, and pyrogen contamination. The tips are available in five volumes and in multiple packaging options, including environmentally friendly reload stacks.
ClipTip® 200 pipette tips suit pipetting of 10 to 200 µl volumes.
The innovative threeMore Product Information