Anti-SLITRK5 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX85414
89329-478EA 980.99 CAD
89329-478
Anti-SLITRK5 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Slitrk5

Type: Primary
Antigen: SLITRK5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol SLITRK5
Environmentally Preferable
Antigen name SLIT And NTRK-like Family member 5
Antigen synonyms bA364G4.2|SLIT and NTRK-like protein 5|Slitrk5|KIAA0918|Leucine-rich repeat-containing protein 11|LRRC11
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 26050
Isotype
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Purification Affinity chromatography purified via peptide column
Storage buffer PBS containing 0.02% sodium azide
Storage temperature Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR