Anti-ELOVL6 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX85403
89329-456EA 980.99 CAD
89329-456
Anti-ELOVL6 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ELOVL6

Type: Primary
Antigen: ELOVL6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol ELOVL6
Environmentally Preferable
Antigen name Elongation of very long chain fatty acids-like 6
Antigen synonyms Fatty acyl-CoA elongase|FACE|Long-chain fatty-acyl elongase|Elongation of very long chain fatty acids protein 6|hELO2|ELOVL6|LCE
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 79071
Isotype
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Purification Affinity chromatography purified via peptide column
Storage buffer PBS containing 0.02% sodium azide
Storage temperature Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR