Anti-BRSK1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX85055
89328-896EA 980.99 CAD
89328-896
Anti-BRSK1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to BRSK1

Type: Primary
Antigen: BRSK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR