Anti-CHRNB1 Mouse Monoclonal Antibody [clone: INN-hCG-45]

Supplier: Genetex
GTX11393
89317-286EA 979.49 CAD
89317-286
Anti-CHRNB1 Mouse Monoclonal Antibody [clone: INN-hCG-45]
Antibodies
Mouse monoclonal antibody [INN-hCG-45] to hCG (holo c3)

Type: Primary
Antigen: CHRNB1
Clonality: Monoclonal
Clone: INN-hCG-45
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR