Anti-GDF2/BMP9 Mouse Monoclonal Antibody [clone: INN-hFSH 100]

Supplier: Genetex
GTX11383
89317-276EA 979.77 CAD
89317-276
Anti-GDF2/BMP9 Mouse Monoclonal Antibody [clone: INN-hFSH 100]
Antibodies
Mouse monoclonal antibody [INN-hFSH 100] to CG alpha

Type: Primary
Antigen: CGA
Clonality: Monoclonal
Clone: INN-hFSH 100
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR