Anti-IgL Mouse Monoclonal Antibody [clone: HP6054]

Supplier: Genetex
GTX17112
89317-118EA 821.3 CAD
89317-118
Anti-IgL Mouse Monoclonal Antibody [clone: HP6054]
Antibodies
IHC-P: Prediluted and ready to use. An incubation period of 30 minutes at room temperature is recommended. Formalin fixed paraffin embedded tissue sections require high temperature antigen unmasking with 10 mM citrate buffer, pH 6.0 prior to immunostaining. IHC-Fr (acetone fixed): Prediluted and ready to use. An incubation period of 30 minutes at room temperature is recommended. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IgL
Clonality: Monoclonal
Clone: HP6054
Conjugation:
Epitope: Light Chain
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR