Anti-IgG Rabbit Polyclonal Antibody (Biotin)

Supplier: Genetex
GTX25753
89314-934EA 446.84 CAD
89314-934
Anti-IgG Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR