Anti-IgG Goat Polyclonal Antibody (Rhodamine)

Supplier: Genetex
GTX26985
89307-096EA 382.12 CAD
89307-096
Anti-IgG Goat Polyclonal Antibody (Rhodamine)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Goat
Isotype:
Reactivity: Donkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR