Anti-IgG Sheep Polyclonal Antibody (Rhodamine)

Supplier: Genetex
GTX26849
89307-068EA 388.19 CAD
89307-068
Anti-IgG Sheep Polyclonal Antibody (Rhodamine)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Sheep
Isotype:
Reactivity: Rat
Order Now

SPECIFICATIONS

Antigen symbol IgG
Antigen name Immunoglobulin G
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Conjugation Rhodamine
Reactivity Rat
Host Sheep
Isotype
ImmunoChemistry Yes
Flow cytometry Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR