Anti-IgG Rabbit Polyclonal Antibody (Rhodamine)

Supplier: Genetex
GTX26768
89307-052EA 388.05 CAD
89307-052
Anti-IgG Rabbit Polyclonal Antibody (Rhodamine)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Rhodamine
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR