Anti-DDX53 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX81413
89301-912EA 911.27 CAD
89301-912
Anti-DDX53 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to DDX53 (C-term)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:50-100FACS: Use at 1:10-50Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: DDX53
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

SPECIFICATIONS

Antigen symbol DDX53
Antigen name DEAD (Asp-Glu-Ala-Asp) box polypeptide 53
Environmentally Preferable
Antigen synonyms Probable ATP-dependent RNA helicase DDX53|DEAD box protein CAGE|CT26|Cancer/testis antigen 26|CAGE|Cancer-associated gene protein|DEAD box protein 53
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Mouse
Host Rabbit
Gene ID 168400
Isotype IgG
Western blot Yes
ELISA Yes
Gel shift Yes
Flow cytometry Yes
Immunogen KLH conjugated synthetic peptide between 602~631 amino acids from the C-terminal region of human DDX53.
Purification Prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis against PBS.
Concentration 0.25mg/ml
Storage buffer PBS with 0.09% (W/V) sodium azide
Storage temperature Store at at 2-8°C for up to 6 months. For long term storage, aliquot and store at -20 °C. Avoid repeated freeze/thaw cycles.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR