Anti-CDIPT (CDP-diacylglycerol--inositol 3-phosphatidyltransferase) Rabbit Polyclonal Antibody

Supplier: Genetex
GTX80599
89300-832EA 1101.66 CAD
89300-832
Anti-CDIPT (CDP-diacylglycerol--inositol 3-phosphatidyltransferase) Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to CDIPT (Internal)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500IHC: Use at 1:50-100FACS: Use at 1:10-50Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: CDIPT (CDP-diacylglycerol--inositol 3-phosphatidyltransferase)
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR