Anti-EFHD2 Goat Polyclonal Antibody
Supplier: Genetex
GTX89662
89297-934EA 979.65 CAD
89297-934
Anti-EFHD2 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to Swiprosin 1 / EFHD2
Approx 28kDa band observed in Human Lymph Node and Tonsil lysates (calculated MW of 26.7kDa according to NP_077305.2). Recommended concentration: 0.1-0.3µg/ml. In paraffin embedded human tonsil stains cells in the proximity of the epithelial criptae, granulocyte neutrophils, macrophages within the germinal centre and in scattered cells (particularly in the interfollicular area and associated with epithelium) that resemble macrophages and/or dendritic cells. (IHC data have been obtained with EB06552 from previous batch.) This antibody doesMore Product Information