Anti-PREX1 Goat Polyclonal Antibody

Supplier: Genetex
GTX88346
89296-258EA 979.65 CAD
89296-258
Anti-PREX1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to PREX1

Approx 150kDa band observed in Human Brain (Temporal Cortex) lysates (calculated MW of 186kDa according to NP_065871.2). Recommended concentration: 1-3µg/ml.

Type: Primary
Antigen: PREX1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Bovine, Dog, Rat
Order Now

SPECIFICATIONS

Antigen symbol PREX1
Environmentally Preferable
Antigen name Phosphatidylinositol-3,4,5-Trisphosphate-Dependent Rac Exchange Factor 1
Antigen synonyms PREX-1|PREX 1|P-REX1
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Bovine,Dog,Rat,Mouse
Host Goat
Gene ID 57580
Isotype IgG
Western blot Yes
ELISA Yes
Immunogen C-ERSNLPTDASTTA
Purification Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
Concentration 0.5mg/ml
Storage buffer Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
Storage temperature Aliquot and store at –20 °C. Minimize freezing and thawing.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR