Anti-CD8A Rat Monoclonal Antibody [clone: 53-6.7]

Supplier: Genetex
GTX80266
89295-886EA 835.95 CAD
89295-886
Anti-CD8A Rat Monoclonal Antibody [clone: 53-6.7]
Antibodies
Rat monoclonal antibody [53-6.7] to CD8 alpha

Recommended Starting Dilutions:FACS: Use at 1.5 µg/mLIHC-Fr: Note: Formaldehyde fixation is not recommended,. Acetone fixation is preferredFunctional: Isolation and depletion of CD8+ cells, blocking of cytotoxicity, inhibition of CD8+ T cell proliferationOptimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: 53-6.7
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR