Anti-IASPP (protein phosphatase 1 Regulatory (inhibitor) Subunit 13 like) Rabbit Polyclonal Antibody

Supplier: Genetex
GTX37659
89294-906EA 990.93 CAD
89294-906
Anti-IASPP (protein phosphatase 1 Regulatory (inhibitor) Subunit 13 like) Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to IASPP

P-ELISA:1:500-1000, WB:1:100-500, IHC-P:1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: IASPP (protein phosphatase 1 Regulatory (inhibitor) Subunit 13 like)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR