Anti-Ly-6A/E Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: D7]

Supplier: Genetex
GTX43420
89293-076EA 924.88 CAD
89293-076
Anti-Ly-6A/E Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: D7]
Antibodies
Rat monoclonal antibody [D7] to Ly-6A/E

Recommended Starting Dilutions: Flow Cytometry: 1/5 - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: Ly-6A/E
Clonality: Monoclonal
Clone: D7
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR