Anti-HSP70 Mouse Monoclonal Antibody [clone: 5A6]

Supplier: Genetex
GTX82827
89287-304EA 996.68 CAD
89287-304
Anti-HSP70 Mouse Monoclonal Antibody [clone: 5A6]
Antibodies
Mouse monoclonal antibody [5A6] to HSP70

Recommended Starting Dilutions: For ELISA: Use at a dilution of 1:10000. For WB: Use at a dilution of 1:500 - 1:2000. For IHC-P: Use at a dilution of 1:200 - 1:1000. For ICC/IF: Use at a dilution of 1:200 - 1:1000. Not yet tested in other applications. Determining optimal working dilutions by titration test.

Type: Primary
Antigen: HSP70
Clonality: Monoclonal
Clone: 5A6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR