Anti-Ly-6B.2 AlloAntigen Rat Monoclonal Antibody [clone: 39266]

Supplier: Genetex
GTX39794
89284-470EA 878.12 CAD
89284-470
Anti-Ly-6B.2 AlloAntigen Rat Monoclonal Antibody [clone: 39266]
Antibodies
Type: Primary
Antigen: Ly-6B.2 AlloAntigen
Clonality: Monoclonal
Clone: 39266
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR