Anti-NDMAR2B Rabbit Polyclonal Antibody

Supplier: Genetex
GTX31084
89283-834EA 1086.77 CAD
89283-834
Anti-NDMAR2B Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to NMDA NR2B Subunit (phospho Tyr1252)

Type: Primary
Antigen: NDMAR2B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR