Anti-MHC Class I H-2B/D/P/Q/W16 Rat Monoclonal Antibody [clone: ER-HR52]

Supplier: Genetex
GTX39707
89280-488EA 1191.6 CAD
89280-488
Anti-MHC Class I H-2B/D/P/Q/W16 Rat Monoclonal Antibody [clone: ER-HR52]
Antibodies
Type: Primary
Antigen: MHC Class I H-2B/D/P/Q/W16
Clonality: Monoclonal
Clone: ER-HR52
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

SPECIFICATIONS

Antigen symbol MHC Class I H-2B/D/P/Q/W16
Antigen name MHC Class I H-2B/D/P/Q/W16
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Clone ER-HR52
Reactivity Mouse
Host Rat
Isotype

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR