Anti-CR2 Mouse Monoclonal Antibody [clone: CA2.1D6]

Supplier: Genetex
GTX43150
89280-280EA 861.04 CAD
89280-280
Anti-CR2 Mouse Monoclonal Antibody [clone: CA2.1D6]
Antibodies
Type: Primary
Antigen: CR2
Clonality: Monoclonal
Clone: CA2.1D6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR