Anti-CRADD Rabbit Polyclonal Antibody

Supplier: Genetex
GTX28425
89279-182EA 1065.43 CAD
89279-182
Anti-CRADD Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CRADD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR