Anti-STK39 Mouse Monoclonal Antibody [clone: 4000]

Supplier: Genetex
GTX83542
89271-892EA 1136.34 CAD
89271-892
Anti-STK39 Mouse Monoclonal Antibody [clone: 4000]
Antibodies
Mouse monoclonal antibody [4E3] to STK39

Recommended Starting Dilutions:WB 1:1000 IHC 1:50 Flow 1:500Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: STK39
Clonality: Monoclonal
Clone: 4000
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR