Anti-UBXN10 Mouse Monoclonal Antibody [clone: 2F6]

Supplier: Genetex
GTX83450
89271-708EA 1137.02 CAD
89271-708
Anti-UBXN10 Mouse Monoclonal Antibody [clone: 2F6]
Antibodies
Mouse monoclonal antibody [2F6] to UBXN10

Recommended Starting Dilutions:WB 1:2000 ICC/IF 1:100 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: UBXN10
Clonality: Monoclonal
Clone: 2F6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR