Anti-NRK2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX46967
89269-998EA 986.33 CAD
89269-998
Anti-NRK2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ITGB1BP3 (Middle)

Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: NRK2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR