Anti-25 OH Vitamin D3 Mouse Monoclonal Antibody [clone: B2530M]

Supplier: Genetex
GTX44515
89263-124EA 991.12 CAD
89263-124
Anti-25 OH Vitamin D3 Mouse Monoclonal Antibody [clone: B2530M]
Antibodies
Type: Primary
Antigen: 25 OH Vitamin D3
Clonality: Monoclonal
Clone: B2530M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: [delete]
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR