Anti-IgG2a Rat Monoclonal Antibody (PE (Phycoerythrin))

Supplier: Genetex
GTX35058
89262-124EA 268.49 CAD
89262-124
Anti-IgG2a Rat Monoclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Secondary
Antigen: IgG2a
Clonality: Monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR