CentriVap® Complete™ Vacuum Concentrator, Labconco®
Supplier: Labconco
7315022 7315023 7315030 7315031 7315032 7315033 7315040 7315041 7315042 7315043 7315060 7315061 7315062 7315063 7315070 7315071 7315072 7315073 7315020 7315021
89239-638EA 39737 CAD
89239-638 89239-640 89239-642 89239-644 89239-646 89239-648 89239-650 89239-652 89239-654 89239-656 89239-658 89239-660 89239-662 89239-664 89239-666 89239-668 89239-670 89239-672 89259-750 89259-752
CentriVap® Complete™ Vacuum Concentrator, Labconco®
Concentrators
The all-in-one CentriVap® Complete Vacuum Concentrator houses every component needed to begin processing multiple samples
Using centrifugal force, vacuum and controlled heat, the CentriVap® Complete speeds concentration of samples in a variety of applications such as molecular biology, proteomics, genomics, cell biology, microbiology, biochemistry, drug discovery, and analytical chemistry. Acid-resistant models are available for corrosive chemical use. Other models feature Heat Boost™, which surrounds the chamber wall to further speed the evaporation of high boiling point solvents.