Paint and Ink Storage Cabinets, SECURALL®
Supplier: Securall Cabinets
P120 P320 P140 P240 P340 P160 P360 P1120 P3120
89237-726EA 3100.75 CAD
89237-726 89237-728 89237-730 89237-732 89237-734 89237-736 89237-738 89237-740 89237-742
Paint and Ink Storage Cabinets, SECURALL®
Furniture Storage Cabinets
Paint storage cabinets for 1 gal. paint cans are available in 20, 40, 60 and up to 120 gal. capacities.
  • Warning label reads "Flammable - Keep Fire Away"
  • Perfect for print shops or auto body repair
Finished in two-part urethane paint.
Standard color is yellow, please contact your Avantor Furniture Specialist for more information.
Certifications: SMaRT Certified; FM Approved.
Order Now
SPECIFICATIONS
Environmentally Preferable Sustainable Materials
Environmental certification SMaRT Certified