Double Polypropylene Bucket Systems, 6.5 Gallon, Contec®
Supplier: Avantor
89237-624CS 2018.5 CAD
89237-624 89237-628
Double Polypropylene Bucket Systems, 6.5 Gallon, Contec®
Mops
The Double Polypropylene Bucket System is a fully autoclavable stainless steel rolling cart with casters, two 6.5 gallon (25 l) polypropylene buckets (gray, red) and a stainless steel drip cap. The durable and lightweight stainless steel caster base is easy to maneuver around any cleanroom.
  • Ideal for use in smaller cleanrooms or controlled areas where protocols require a two-bucket system
  • Compatible with flat and curved Contec® mops
  • Fully autoclavable
  • Compatible with most disinfectants
The units are available with standard or heavy-duty casters. TheMore Product Information