VWR® Stackable Freezer Boxes
Supplier: VWR International
89235-996 89235-998 89236-000 89236-002 89236-004 89236-006 89236-008 89236-010 89236-012 89236-014 89236-016 89236-018 89236-020 89236-024 89236-026 89236-028 89236-030
89235-996CS 929.85 CAD
89235-996 89235-998 89236-000 89236-002 89236-004 89236-006 89236-008 89236-010 89236-012 89236-014 89236-016 89236-018 89236-020 89236-024 89236-026 89236-028 89236-030
VWR® Stackable Freezer Boxes
Boxes Cooling/Freezing Boxes and Bags
VWR® Stackable Freezer Boxes are suitable to store in temperatures as low as –86 °C.
  • Made with PP material
  • Base made with color options natural, blue, green, purple, and orange
  • Lid has grid option for coding
Boxes are stackable. Designed for 50, 81, or 100 places to store both internal and external threaded vials (1 to 2 ml).
Slot width: 12.5 mm (±0.3 mm)