SP Bel-Art Grande Desiccator Cabinets, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F43000-1010 F43010-1010 F43020-1010
89234-884EA 5867.46 CAD
89234-884 89234-886 89234-888
SP Bel-Art Grande Desiccator Cabinets, Bel-Art Products, a part of SP
Desiccators Desiccator Cabinets
Extra-large Scienceware® desiccator cabinets provide a low-humidity, dust-free environment for several large containers or a lot more small ones.
  • Copolyester side and top panel construction
  • Polycarbonate door and rear panel
  • Perforate acrylic shelves promote air circulation
  • Dust-free, low-humidity environment
Clear on all sides for quick visualization of desiccator contents, they are constructed of rugged copolyester side and top panels and a polycarbonate door and rear panel.
Pull-out, perforated acrylic shelves (2 included) allow maximum airMore Product Information