Puritan® UniTranz-RT® Media Transport Systems, Puritan Medical Products
Supplier: Puritan Medical Products
UT-106 UT-116 UT-117 UT-306 UT-316 UT-317 UT-367 UT-302 UT-366 UT-361
89233-434CS 1178.04 CAD
89233-434 89233-436 89233-438 89233-440 89233-442 89233-444 89233-446 89233-450 89233-452 89233-454
Puritan® UniTranz-RT® Media Transport Systems, Puritan Medical Products
Swabs Transport Swabs
Puritan® UniTranz-RT® Transport Systems are used for the collection and preservation of Virus, Chlamydia, Mycoplasma, and Ureaplasma specimens.
Ready-to-use UniTranz-RT® Transport Systems are filled Universal Transport Medium, which is stable at room temperature. Self-centering cap is leak-proof and facilitates easy swab removal and disposal.
The patented PurFlock Ultra® or spun polyester swabs come in a variety of tip shapes and sizes which enhance collection flexibility and improve patient comfort. Made in the USA.
Certifications: CE marked.
Ordering information: Products are final sale.
Order Now
SPECIFICATIONS
Environmentally Preferable