ToxinEraser™ Endotoxin Resin Removal Kit, Genscript
Supplier: Genscript
L00338
89233-330EA 213.84 CAD
89233-330
ToxinEraser™ Endotoxin Resin Removal Kit, Genscript
Cleaning Agents Decontaminants
The ToxinEraser™ endotoxin removal resin utilizes immobilized polymixin B to bind and remove pyrogens from solution.
  • High stability and removal efficiency
  • Excellent binding capacity of over 2000000 EU/mL (CV)
  • Strong resilience
The polymixins are a family of antibiotics that contain a cationic cyclopeptide with a fatty acid chain. Polymixin B neutralizes the biological activity of endotoxins by binding to the lipid A portion of bacterial lipopolysaccharide.