SilTite® μ-Unions, Gas Chromatography Connections, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
073560 073561 073562 073563 073564 073565 073566 073567 073568 073569 073570 073571 073572 073573 073574
89232-106EA 385.78 CAD
89232-106 89232-108 89232-110 89232-112 89232-114 89232-116 89232-118 89232-120 89232-122 89232-124 89232-126 89232-128 89232-130 89232-132 89232-134
SilTite® μ-Unions, Gas Chromatography Connections, Trajan Scientific and Medical
Fittings and Connectors
Your chromatography analysis does not end with the selection of the GC column
  • Inert
  • Zero dead volume
  • Optimized peak shapes
  • Robust and stable
  • For both extreme temperature and pressure cycling
  • Easy to install
  • Leak-free
  • No need to re-tighten after installation
  • Only 9 mm in total length
  • Mass of <0.5 g
  • Low thermal mass
These micro unions feature chemically deactivated stainless steel ferrules alleviating active sites. They should be used when fitting a pre-column or guard column, GC to GC connections.
The combination of components selected for yourMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable