Neptune® S³ Low Retention, Extended Length Pipette Tips
Supplier: Biotix
63300553 63300558 63300557
89230-228CS 1268.84 CAD
89230-228 89230-230 89230-232
Neptune® S³ Low Retention, Extended Length Pipette Tips
Pipette Tips
The exclusive S³ polymer delivers to within 0.1% of indicated volume, eliminating sample hold-up and ensuring a more precise application.
Eco-friendly empty pipette tip trays are manufactured from recycled polypropylene. The trays are autoclavable, enabling reuse for maximum sustainability. When combined with ESP™ reload systems, these tip trays reduce plastic waste by up to 90%. The trays provide superior stability, firm placement, and a maximum space saving design.
This proprietary technology is essential for quantitative assays where sample retention can hinder accurate results.More Product Information