VWR® Gold-N-Gel™ RNA dye 200X, Biotechnology Grade
Supplier: VWR
N734-5ML
89230-108EA 152.19 CAD
89230-108
VWR® Gold-N-Gel™ RNA dye 200X, Biotechnology Grade
Stains Electrophoresis Stains
Supplied as a 200X concentrate, Gold-N-Gel™ RNA Stain is a non-mutagenic, fluorescent in-gel stain for immediate visualization of RNA bands on denaturing agarose gels.
  • Safer than ethidium bromide
  • Immediate band visualization with UV illumination - no post-run staining or destaining
  • Easy-to-use
Gold-N-Gel™ RNA Stain is compatible with most standard protocols for denaturing electrophoresis of RNA fragments. Bands are immediately visible upon illumination with UV light. A standard ethidium bromide filter can be used for gel documentation. Available in 12.5mL solution and in 5 or 15mL dropper bottle sizes.
Specifications
Abs.@ Lambda Max: 0.6 - 0.7
Lambda Max: 338 - 345 nm
RNase: NONE / Fail