Valve Port Hat-Type Specialty Needles, Hamilton Company
Supplier: Hamilton
90815 90817 90816 90818 90819 90821 90822 90823 90824 90825 90826 90820
65515-061PK 212.06 CAD
65515-061 65515-141 89221-526 89221-528 89221-530 89221-534 89221-536 89221-538 89221-540 89221-542 89221-544 89221-532
Valve Port Hat-Type Specialty Needles, Hamilton Company
Syringe Needles
Hamilton Company offers the most comprehensive selection of standard and custom needles in the industry
Hat-type needles are designed to be connected directly to a flat bottom valve port. Use the Hamilton Needle Bushing part number 35056 to connect the needle directly to any 1/4-28 Hamilton valve. Custom Length and Gauge
Each specialty needle is made to order. Custom lengths range from 0.375 to 12 inches. Available point styles include 2, 3 and 4. Please specify the desired length and point style when ordering.
Needle Point Styles (Custom lengths and point styles available)
Point Style 2More Product Information