Chemically Inert HV Standard Manual Valves, Hamilton Company
Supplier: Hamilton
86777 86778 86788 86725 86727 86728 86731 86735 86779 86905 86915 86730 86732 86736 86737 86738 86775 86780 86785 86786 86787 86901 86903 86904 86907 86910 86911 86912 86913 86916 86918 86726 86729 86781
65510-590EA 277.76 CAD
65510-590 65510-636 65510-953 65510-035 65510-115 65510-159 65510-272 65510-352 65510-670 65511-195 65512-700 89221-322 89221-324 89221-326 89221-328 89221-330 89221-332 89221-336 89221-340 89221-342 89221-344 89221-346 89221-348 89221-350 89221-354 89221-356 89221-358 89221-360 89221-362 89221-364 89221-366 89221-368 89221-318 89221-320 89221-334 89221-338
Chemically Inert HV Standard Manual Valves, Hamilton Company
Valves Vacuum Valves
Hamilton Company offers the most comprehensive selection of standard and custom syringes, valves and accessories in the industry
Each is manufactured to achieve the highest level of accuracy and precision possible. Hamilton (HV) Plug Valves provide an economical and flexible method of custom-building a fluid flow control system, from a simple on/off valve to a more complex multi-valve arrangement, using a combination of loop and distribution valves. The small valve body with its chemically inert flow path is ideally suited for low pressure applications. All HV Plug Valves allow you toMore Product Information