Microliter™ 700, Gastight® 1700, Microliter™ 7000 Series Digital Syringes, Hamilton
Supplier: Hamilton
DS80530 DS80600 DS80700 DS86259 DS86250 DS86200 DS88500 DS80075 DS80030 DS80230 DS81030 DS81230 DS81175 DS87930 DS80000 DS80100 DS80135 DS80200 DS80275 DS80300 DS80330 DS80365 DS80383 DS80400 DS80430 DS80465 DS80500 DS80565 DS80630 DS80665 DS80730 DS80765 DS80800 DS80830 DS80865 DS80900 DS80930 DS80975 DS81000 DS81075 DS81100 DS81130 DS81217 DS86211 DS87900 DS88000 DS88011 DS88411 DS88511 DS88400
25501-042EA 4430.27 CAD
25501-042 25501-044 25501-050 25501-062 25501-064 25501-072 25501-080 25501-092 25501-094 25501-112 25501-124 25501-134 25501-136 89210-106 89221-180 89221-182 89221-184 89221-186 89221-188 89221-190 89221-192 89221-194 89221-196 89221-198 89221-200 89221-202 89221-204 89221-206 89221-208 89221-210 89221-212 89221-214 89221-216 89221-218 89221-220 89221-222 89221-224 89221-226 89221-228 89221-230 89221-232 89221-234 89221-236 89221-238 89221-240 89221-242 89221-244 89221-248 89221-250 89221-246
Microliter™ 700, Gastight® 1700, Microliter™ 7000 Series Digital Syringes, Hamilton
Syringes Syringes for Chromatography
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry. Each is manufactured to achieve the highest level of accuracy and precision possible.
Reading graduations is always a challenge
Hamilton digital syringes eliminate parallax errors by providing a digital readout on an integrated LCD screen. An adjustable plunger-stop guarantees reproducible plunger placement. Easily achieve precision up to 0.5% while dispensing volumes from 500 µL down to 50 nL. Easy-To-Read
Now you don't have to read between the lines to determine how much sample isMore Product Information