Cole-Parmer® SI-200 Series Stuart® Microplate Shaking Incubators, Antylia Scientific
Supplier: Antyila Scientific
79520-08
89218-350EA 8117.17 CAD
89218-350
Cole-Parmer® SI-200 Series Stuart® Microplate Shaking Incubators, Antylia Scientific
Shakers Shaking Incubators
Specifically designed to mix small volumes of liquid in a temperature-controlled environment.
  • Microprocessor controlled shaking speed ensures accuracy
  • High shaking speed up to 1250 rpm
  • 1.5 mm diameter orbit is ideal for microplates and microtubes
  • Easily set and view temperature and speed on the digital display
  • Unique retractable platform offers easy loading and unloading
These microplate shaking incubators combine high speed with tiny orbit to create the vibrational shaking action required for small sample volumes. All models feature a timer which canMore Product Information